Merck
  • Multivalency Beats Complexity: A Study on the Cell Uptake of Carbohydrate Functionalized Nanocarriers to Dendritic Cells.
Multivalency Beats Complexity: A Study on the Cell Uptake of Carbohydrate Functionalized Nanocarriers to Dendritic Cells.
Cells (2020-09-17)
Matthias Krumb, Marie-Luise Frey, Jens Langhanki, Robert Forster, Danuta Kowalczyk, Volker Mailänder, Katharina Landfester, Till Opatz
ABSTRACT
Herein, we report the synthesis of carbohydrate and glycodendron structures for dendritic cell targeting, which were subsequently bound to hydroxyethyl starch (HES) nanocapsules prepared by the inverse miniemulsion technique. The uptake of the carbohydrate-functionalized HES nanocapsules into immature human dendritic cells (hDCs) revealed a strong dependence on the used carbohydrate. A multivalent mannose-terminated dendron was found to be far superior in uptake compared to the structurally more complex oligosaccharides used.
Sigma-Aldrich
Dibenzocyclooctyne-PEG4-N-hydroxysuccinimidyl ester, ≥90%