Merck
  • Decreased lymphatic HIF-2α accentuates lymphatic remodeling in lymphedema.
Decreased lymphatic HIF-2α accentuates lymphatic remodeling in lymphedema.
The Journal of clinical investigation (2020-07-17)
Xinguo Jiang, Wen Tian, Eric J Granucci, Allen B Tu, Dongeon Kim, Petra Dahms, Shravani Pasupneti, Gongyong Peng, Yesl Kim, Amber H Lim, F Hernan Espinoza, Matthew Cribb, J Brandon Dixon, Stanley G Rockson, Gregg L Semenza, Mark R Nicolls
ABSTRACT
Pathologic lymphatic remodeling in lymphedema evolves during periods of tissue inflammation and hypoxia through poorly defined processes. In human and mouse lymphedema, there is a significant increase of hypoxia inducible factor 1 α (HIF-1α), but a reduction of HIF-2α protein expression in lymphatic endothelial cells (LECs). We questioned whether dysregulated expression of these transcription factors contributes to disease pathogenesis and found that LEC-specific deletion of Hif2α exacerbated lymphedema pathology. Even without lymphatic vascular injury, the loss of LEC-specific Hif2α caused anatomic pathology and a functional decline in fetal and adult mice. These findings suggest that HIF-2α is an important mediator of lymphatic health. HIF-2α promoted protective phosphorylated TIE2 (p-TIE2) signaling in LECs, a process also replicated by upregulating TIE2 signaling through adenovirus-mediated angiopoietin-1 (Angpt1) gene therapy. Our study suggests that HIF-2α normally promotes healthy lymphatic homeostasis and raises the exciting possibility that restoring HIF-2α pathways in lymphedema could mitigate long-term pathology and disability.
Sigma-Aldrich
Human Dermal Lymphatic Endothelial Cells (HDLEC), Adult, 500,000 cryopreserved cells
Sigma-Aldrich
Tamoxifen, ≥99%