Merck
  • Metabotropic glutamate type 5 receptor binding availability during dextroamphetamine sensitization in mice and humans.
Metabotropic glutamate type 5 receptor binding availability during dextroamphetamine sensitization in mice and humans.
Journal of psychiatry & neuroscience : JPN (2020-06-20)
Kelly Smart, Atsuko Nagano-Saito, Michele S Milella, Diana Yae Sakae, Mathieu Favier, Erika Vigneault, Leanne Louie, Alison Hamilton, Stephen S G Ferguson, Pedro Rosa-Neto, Sridar Narayanan, Salah El Mestikawy, Marco Leyton, Chawki Benkelfat
ABSTRACT
Glutamate transmission is implicated in drug-induced behavioural sensitization and the associated long-lasting increases in mesolimbic output. Metabotropic glutamate type 5 (mGlu5) receptors might be particularly important, but most details are poorly understood. We first assessed in mice (n = 51, all male) the effects of repeated dextroamphetamine administration (2.0 mg/kg, i.p.) on locomotor activity and binding of the mGlu5 ligand [3H]ABP688. In a parallel study, in 19 stimulant-drug-naïve healthy human volunteers (14 female) we administered 3 doses of dextroamphetamine (0.3 mg/kg, p.o.) or placebo, followed by a fourth dose 2 weeks later. We measured [11C]ABP688 binding using positron emission tomography before and after the induction phase. We assessed psychomotor and behavioural sensitization using speech rate, eye blink rate and self-report. We measured the localization of mGlu5 relative to synaptic markers in mouse striatum using immunofluorescence. We observed amphetamine-induced psychomotor sensitization in mice and humans. We did not see group differences in mGlu5 availability following 3 pre-challenge amphetamine doses, but group differences did develop in mice administered 5 doses. In mice and humans, individual differences in mGlu5 binding after repeated amphetamine administration were negatively correlated with the extent of behavioural sensitization. In drug-naïve mice, mGlu5 was expressed at 67% of excitatory synapses on dendrites of striatal medium spiny neur. Correlational results should be interpreted as suggestive because of the limited sample size. We did not assess sex differences. Together, these results suggest that changes in mGlu5 availability are not part of the earliest neural adaptations in stimulant-induced behavioural sensitization, but low mGlu5 binding might identify a higher propensity for sensitization.
Sigma-Aldrich
Anti-mGluR5 Antibody, clone 11D9.1, clone 11D9.1, from mouse
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®