Merck
  • BMP2K dysregulation promotes abnormal megakaryopoiesis in acute megakaryoblastic leukemia.
BMP2K dysregulation promotes abnormal megakaryopoiesis in acute megakaryoblastic leukemia.
Cell & bioscience (2020-04-24)
Manman Wang, Tan Zhang, Xuechun Zhang, Zhou Jiang, Min Peng, Zan Huang
ABSTRACT
Forced polyploidization is an effective strategy for acute megakaryoblastic leukemia (AMKL) therapy and factors controlling polyploidization are potential targets for drug development. Although bone morphology protein 2-inducible kinase (BMP2K) has been implied to be a potential target for fasudil, a potent polyploidy-inducing compound, the function of BMP2K in megakaryopoiesis and AMKL remains unknown. This study aimed to investigate the role of BMP2K as a novel regulator in megakaryocyte polyploidization and differentiation and its implication in AMKL therapy. BMP2K upregulation was observed in human megakaryopoiesis and leukemia cells whereas BMP2K was downregulated in AMKL cells forced to undergo terminal differentiation. Functionally, BMP2K suppressed MLN8237-induced megakaryocytic differentiation in AMKL cells and dampened megakaryocyte differentiation in primary mouse fetal liver cells. Furthermore, BMP2K overexpression conferred resistance to multiple chemotherapy compounds in AMKL cells. Mechanistically, cyclin-dependent kinase 2 (CDK2) interacted with BMP2K and partially mediated its function. In transient MLN8237 and nocodazole challenge cell model, BMP2K reduced cell percentage of G2/M phase but increased G1 phase, suggesting a role of BMP2K antagonizing polyploidization and promoting mitosis by regulating cell cycle in megakaryopoiesis. BMP2K negatively regulates polyploidization and megakaryocyte differentiation by interacting CDK2 and promoting mitosis in megakaryopoiesis. BMP2K may serve as a potential target for improvement of AMKL therapy.
Sigma-Aldrich
K03861, ≥98% (HPLC)