Merck
Quantifying CDK inhibitor selectivity in live cells.
Nature communications (2020-06-04)
Carrow I Wells, James D Vasta, Cesear R Corona, Jennifer Wilkinson, Chad A Zimprich, Morgan R Ingold, Julie E Pickett, David H Drewry, Kathryn M Pugh, Marie K Schwinn, Byounghoon Brian Hwang, Hicham Zegzouti, Kilian V M Huber, Mei Cong, Poncho L Meisenheimer, Timothy M Willson, Matthew B Robers
ABSTRACT
Concerted multidisciplinary efforts have led to the development of Cyclin-Dependent Kinase inhibitors (CDKi's) as small molecule drugs and chemical probes of intracellular CDK function. However, conflicting data has been reported on the inhibitory potency of CDKi's and a systematic characterization of affinity and selectivity against intracellular CDKs is lacking. We have developed a panel of cell-permeable energy transfer probes to quantify target occupancy for all 21 human CDKs in live cells, and present a comprehensive evaluation of intracellular isozyme potency and selectivity for a collection of 46 clinically-advanced CDKi's and tool molecules. We observed unexpected intracellular activity profiles for a number of CDKi's, offering avenues for repurposing of highly potent molecules as probes for previously unreported targets. Overall, we provide a broadly applicable method for evaluating the selectivity of CDK inhibitors in living cells, and present a refined set of tool molecules to study CDK function.
Sigma-Aldrich
K03861, ≥98% (HPLC)