Merck
  • Impact of phenol on the glycerophospholipid turnover and potential role of circadian clock in the plant response against this pollutant in tobacco hairy roots.
Impact of phenol on the glycerophospholipid turnover and potential role of circadian clock in the plant response against this pollutant in tobacco hairy roots.
Plant physiology and biochemistry : PPB (2020-04-14)
Lucas G Sosa Alderete, Sabrina Flor, Silvia Lucangioli, Elizabeth Agostini
ABSTRACT
Glycerophospholipids (GPLs) from cell membranes (CM) are a proper source for the synthesis of lipid messengers able to activate signal pathways that will define the plant survival under changing and stressful environmental conditions. Little is known about how GPLs metabolism (GPLsM) is regulated and the effects of phenol treatment on GPLs composition. In this work, we studied the effects of phenol both on GPLs turnover and on the expression of GPLsM-related genes potentially regulated by the circadian clock, using tobacco hairy root cultures (HRC). Phenol decreased the total PC levels and increased PE, PG and CL levels in the dark phase. Different molecular species of PC and PE showed the same trend than the total PC and PE upon phenol treatment. Besides, significant differences in the expression of all studied genes related to GPLsM were found. NtCCT2 expression was affected at all analyzed times while NtPECT1 and NtAAPT1 showed similar expression patterns. NtCDS1, NtPGPS2 and NtCLS genes showed significant and differential expression profiles both in untreated and treated HRC. PECT1 and NtPGPS2 genes seem to conserve a circadian expression profile mainly in untreated HRC. However, phenol was able to modify the GPLs composition and the expression of genes related to GPLs synthesis. The GPLs modification could be explained by the up-regulation of NtPECT1, NtAAPT1 and NtCLS genes during the dark phase, suggesting for being a crucial moment for HRC to trigger an adaptive response against this organic pollutant.