Merck
  • A potential strategy for in-stent restenosis: Inhibition of migration and proliferation of vascular smooth muscle cells by Cu ion.
A potential strategy for in-stent restenosis: Inhibition of migration and proliferation of vascular smooth muscle cells by Cu ion.
Materials science & engineering. C, Materials for biological applications (2020-07-01)
Yuan Zhang, Xiaoyan Wang, Zhiling Ma, Bing Bai, Jie Liu, Lei Yang, Gaowu Qin, Erlin Zhang
ABSTRACT
The in-stent restenosis (ISR) often happens after the implantation of metal stents, including both bare metal stents (BMSs) and drug-eluting stents (DESs). Drug release from DESs could reduce significantly the occurrence of ISR but also suppress the revascularization and cause thrombosis. In this study, the effect of Cu ion in a range of 0 to 500 μM on the migration and proliferation of rat aortic smooth muscle cells (RASMCs) was investigated by a series of in vitro experiments including wound-healing assay, cell viability assay and flow cytometric analysis. It has been found that the critical concentration of Cu ion should be at least 250 μM in order to significantly inhibit the migration of RASMCs and the proliferation of RASMCs were impeded by every dose of Cu ion used in this study. In addition, the protein level of caspase-3 was upregulated by 250 μM and 500 μM Cu2+ exposure, which might be the main reason for RASMCs apoptosis. Thus, it is proposed that ISR might be prevented by the constant release of Cu ion.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Rat Smooth Muscle Cell Growth Medium (500 ml)