Merck
  • Installation of Minimal Tetrazines through Silver-Mediated Liebeskind-Srogl Coupling with Arylboronic Acids.
Installation of Minimal Tetrazines through Silver-Mediated Liebeskind-Srogl Coupling with Arylboronic Acids.
Journal of the American Chemical Society (2019-10-12)
William D Lambert, Yinzhi Fang, Subham Mahapatra, Zhen Huang, Christopher W Am Ende, Joseph M Fox
ABSTRACT
Described is a general method for the installation of a minimal 6-methyltetrazin-3-yl group via the first example of a Ag-mediated Liebeskind-Srogl cross-coupling. The attachment of bioorthogonal tetrazines on complex molecules typically relies on linkers that can negatively impact the physiochemical properties of conjugates. Cross-coupling with arylboronic acids and a new reagent, 3-((p-biphenyl-4-ylmethyl)thio)-6-methyltetrazine (b-Tz), proceeds under mild, PdCl2(dppf)-catalyzed conditions to introduce minimal, linker-free tetrazine functionality. Safety considerations guided our design of b-Tz which can be prepared on decagram scale without handling hydrazine and without forming volatile, high-nitrogen tetrazine byproducts. Replacing conventional Cu(I) salts used in Liebeskind-Srogl cross-coupling with a Ag2O mediator resulted in higher yields across a broad library of aryl and heteroaryl boronic acids and provides improved access to a fluorogenic tetrazine-BODIPY conjugate. A covalent probe for MAGL incorporating 6-methyltetrazinyl functionality was synthesized in high yield and labeled endogenous MAGL in live cells. This new Ag-mediated cross-coupling method using b-Tz is anticipated to find additional applications for directly introducing the tetrazine subunit to complex substrates.
Sigma-Aldrich
3-(([1,1′-Biphenyl]-4-ylmethyl)thio)-6-methyl-1,2,4,5-tetrazine, >98%