Merck
Properties of diphytanoyl phospholipids at the air-water interface.
Langmuir : the ACS journal of surfaces and colloids (2014-12-05)
Anthony Yasmann, Sergei Sukharev
ABSTRACT
Diphytanoylphosphatidyl choline (DPhPC) is a synthetic ester lipid with methylated tails found in archaeal ether lipids. Because of the stability of DPhPC bilayers and the absence of phase transitions over a broad range of temperatures, the lipid is used as an artificial membrane matrix for the reconstitution of channels, pumps, and membrane-active peptides. We characterized monomolecular films made of DPhPC and its natural ether analog DOPhPC at the air-water interface. We measured compression isotherms and dipole potentials of films made of DPhPC, DPhPE, and DOPhPC. We determined that at 40 mN/m the molecular area of DPhPC is 81.2 Å(2), consistent with X-ray and neutron scattering data obtained in liposomes. This indicates that 40 mN/m is the monolayer-bilayer equivalence pressure for this lipid. At this packing density, the compressibility modulus (Cs(-1 )= 122 ± 7 mN/m) and interfacial dipole potential (V = 355 ± 16 mV) were near their maximums. The molecular dipole moment was estimated to be 0.64 ± 0.02 D. The ether DOPhPC compacted to 70.4 Å(2)/lipid at 40 mN/m displaying a peak compressibility similar to that of DPhPC. The maximal dipole potential of the ether lipid was about half of that for DPhPC at this density, and the elemental dipole moment was about a quarter. The spreading of DPhPC and DOPhPC liposomes reduced the surface tension of the aqueous phase by 46 and 49 mN/m, respectively. This corresponds well to the monolayer collapse pressure. The equilibration time shortened as the temperature increased from 20 to 60 °C, but the surface pressure at equilibrium did not change. The data illustrates the properties of branched chains and the contributions of ester bonds in setting the mechanical and electrostatic parameters of diphytanoyl lipids. These properties determine an environment in which reconstituted voltage- or mechano-activated proteins may function. Electrostatic properties are important in the preparation of asymmetric folded bilayers, whereas lateral compressibility defines the tension in mechanically stimulated droplet interface bilayers.
Product Description
Avanti
4ME 16:0 PE, 1,2-diphytanoyl-sn-glycero-3-phosphoethanolamine, powder