Merck
New insights on the asymmetric hydroboration of perfluoroalkenes.
Chemical communications (Cambridge, England) (2004-02-07)
Anna M Segarra, Carmen Claver, Elena Fernandez
ABSTRACT
Enantioselective access to Markovnikov regioisomeric perfluoroalcohols is achieved in the presence of chiral cationic rhodium complexes and specific hydroborating reagents.