- The effect of geometric isomerism on the anticancer activity of the monofunctional platinum complex trans-[Pt(NH3)2(phenanthridine)Cl]NO3.
The effect of geometric isomerism on the anticancer activity of the monofunctional platinum complex trans-[Pt(NH3)2(phenanthridine)Cl]NO3.
Chemical communications (Cambridge, England) (2018-02-28)
Wen Zhou, Mohammad Almeqdadi, Michael E Xifaras, Imogen A Riddell, Ömer H Yilmaz, Stephen J Lippard
PMID29484327
ABSTRACT
A trans-DDP based monofunctional phenanthridine Pt(ii) complex was synthesized and characterized. Its anticancer activity was studied in vitro on a panel of human cancer cell lines and mouse intestinal cancer organoids. This complex displays significant antitumor properties, with a different spectrum of activity than that of classic bifunctional cross-linking agents like cisplatin.
MATERIALS