Merck
PTPσ inhibitors promote hematopoietic stem cell regeneration.
Nature communications (2019-08-16)
Yurun Zhang, Martina Roos, Heather Himburg, Christina M Termini, Mamle Quarmyne, Michelle Li, Liman Zhao, Jenny Kan, Tiancheng Fang, Xiao Yan, Katherine Pohl, Emelyne Diers, Hyo Jin Gim, Robert Damoiseaux, Julian Whitelegge, William McBride, Michael E Jung, John P Chute
ABSTRACT
Receptor type protein tyrosine phosphatase-sigma (PTPσ) is primarily expressed by adult neurons and regulates neural regeneration. We recently discovered that PTPσ is also expressed by hematopoietic stem cells (HSCs). Here, we describe small molecule inhibitors of PTPσ that promote HSC regeneration in vivo. Systemic administration of the PTPσ inhibitor, DJ001, or its analog, to irradiated mice promotes HSC regeneration, accelerates hematologic recovery, and improves survival. Similarly, DJ001 administration accelerates hematologic recovery in mice treated with 5-fluorouracil chemotherapy. DJ001 displays high specificity for PTPσ and antagonizes PTPσ via unique non-competitive, allosteric binding. Mechanistically, DJ001 suppresses radiation-induced HSC apoptosis via activation of the RhoGTPase, RAC1, and induction of BCL-XL. Furthermore, treatment of irradiated human HSCs with DJ001 promotes the regeneration of human HSCs capable of multilineage in vivo repopulation. These studies demonstrate the therapeutic potential of selective, small-molecule PTPσ inhibitors for human hematopoietic regeneration.
Sigma-Aldrich
DJ001 (E/Z), ≥98% (HPLC)
Pricing and availability is not currently available.