Merck
  • Determination of free medium-chain fatty acids in beer by stir bar sorptive extraction.
Determination of free medium-chain fatty acids in beer by stir bar sorptive extraction.
Journal of chromatography. A (2008-05-30)
Tomás Horák, Jirí Culík, Marie Jurková, Pavel Cejka, Vladimír Kellner
ABSTRACT
Free medium-chain fatty acids in beer originate from raw materials, mainly from the fermentation activity of yeasts, and can influence beer taste, vitality of yeasts and also the foam stability of beer. This study presents the development of the method for the determination of free fatty medium-chain acids including caproic acid, caprylic acid, capric acid and lauric acid in beer or wort using stir bar sorptive extraction (SBSE). The combination of this extraction technique with solvent back extraction of the extracted analytes and subsequent gas chromatographic analysis with flame ionization detection was used for the determination of these compounds. The influences of different solvent back solutions, sampling time, solvent back extraction times and different contents of ethanol were studied. The method had high repeatability (RSD <6.7%), good linearity (the correlation coefficients were higher than 0.9963 for quadratic curves over the concentration range 0.5-8.0mg/l) and recoveries 57-89%.