Merck
  • Knockdown of TFAM in Tumor Cells Retarded Autophagic Flux through Regulating p53 Acetylation and PISD Expression.
Knockdown of TFAM in Tumor Cells Retarded Autophagic Flux through Regulating p53 Acetylation and PISD Expression.
Cancers (2020-02-26)
Xu Jiang, Jun Wang
ABSTRACT
Mitochondrial transcription factor A (TFAM) is required for mitochondrial DNA replication and transcription, which are essential for mitochondrial biogenesis. Previous studies reported that depleting mitochondrial functions by genetic deletion of TFAM impaired autophagic activities. However, the underlying mechanisms remain largely unknown. In the current study, we identified that knockdown of TFAM repressed the synthesis of autophagy bio-marker LC3-II in tumor cells and decreased the expression of phosphatidyl-serine decarboxylase (PISD). Besides, downregulation of PISD with siRNA reduced the level of LC3-II, indicating that depletion of TFAM retarded autophagy via inhibiting PISD expression. Furthermore, it was found that the tumor repressor p53 could stimulate the transcription and expression of PISD by binding the PISD enhancer. Additionally, the protein stability and transcriptional activity of p53 in TFAM knockdown tumor cells was attenuated, and this was associated with decreased acetylation, especially the acetylation of lysine 382 of p53. Finally, we identified that TFAM knockdown increased the NAD+/NADH ratio in tumor cells. This led to the upregulation of Sirtuin1 (SIRT1), a NAD-dependent protein deacetylase, to deacetylate p53 and attenuated its transcriptional activation on PISD. In summary, our study discovered a new mechanism regarding disturbed autophagy in tumor cells with mitochondrial dysfunction due to the depletion of TFAM.
Sigma-Aldrich
MISSION® esiRNA, targeting human TFAM