Merck
  • Solid-phase microextraction coatings based on the metal-organic framework ZIF-8: Ensuring stable and reusable fibers.
Solid-phase microextraction coatings based on the metal-organic framework ZIF-8: Ensuring stable and reusable fibers.
Talanta (2020-04-22)
Priscilla Rocío-Bautista, Adrián Gutiérrez-Serpa, Alexander John Cruz, Rob Ameloot, Juan H Ayala, Ana M Afonso, Jorge Pasán, Sabina Rodríguez-Hermida, Verónica Pino
ABSTRACT
Chemical vapor deposition of MOFs (MOF-CVD) has been used to coat solid-phase microextraction (SPME) fibers with ZIF-8, by exposing ZnO layers to the linker vapor (2-methylimidazole). This ZIF-8 coating has been used as a seed layer in a following solvothermal MOF growth step in order to increase the ZIF-8 thickness. The combined MOF-CVD and solvothermal growth of ZIF-8 on the fibers result in a thickness of ~3 μm, with adequate thermal stability, and mechanical integrity when tested with methanol and acetonitrile ultrasonic treatments. The fibers have been evaluated in direct immersion mode using gas chromatography and flame ionization detection (GC-FID), for a group of target analytes including three polycyclic aromatic hydrocarbons (PAHs) and five personal care products (PCPs). The optimized conditions of the SPME-GC-FID methods include low amount of aqueous sample (5 mL), stirring for 45 min at 35 °C, and desorption at 280 °C for 5 min. The method presents limits of detection down to 0.6 μg L-1; intra-day, inter-day and inter-batch relative standard deviation values lower than 16%, 19%, and 23%, respectively; and a lifetime higher than 70 cycles.
Sigma-Aldrich
Sodium formate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Pricing and availability is not currently available.