Merck
  • Protective effect of Spirulina platensis against physiological, ultrastructural and cell proliferation damage induced by furan in kidney and liver of rat.
Protective effect of Spirulina platensis against physiological, ultrastructural and cell proliferation damage induced by furan in kidney and liver of rat.
Ecotoxicology and environmental safety (2020-02-06)
Samah R Khalil, Hoda F A Salem, Mohamed M M Metwally, Rasha M Emad, Khlood M Elbohi, Sozan A Ali
ABSTRACT
The modulatory role of the Spirulina platensis (SP) against furan-induced (FU) hepatic and renal damage was assessed in this study. For achieving this, sixty rats were distributed into six groups: control group, SP-administered group (300 mg/kg b.wt orally for 28 days), a FU-intoxicated group (16 mg/kg b.wt, orally, daily for 28 days), protective co-treated group SP/F (administered SP 300 mg/kg b.wt, one week before, and concurrently with FU intoxication), therapeutic co-treated group FU/SP (administered SP 300 mg/kg b.wt, one week after FU intoxication for 28 days) and protective/therapeutic co-treated group SP/FU/SP (administered SP one week before and after, concurrently with FU intoxication). Subsequently, the biochemical responses and the histology of hepatic and renal tissues in treated rats were assessed. The results indicated that FU intoxication induced a significant hepato- and nephropathy represented by the elevation in the values of tissue injury biomarkers and reduction in protein levels. Histologically, a wide range of morphological, cytotoxic, inflammatory, and vascular alterations as well as downregulation in the immunoexpression of the proliferating cell nuclear antigen (PCNA) and the proliferation-associated nuclear antigen (Ki-67) were induced by FU intoxication. Oral SP administration, particularly in the protective/therapeutic co-treated group, markedly supressed the serum levels of the tissue injury biomarkers, diminished the inflammatory response, restored the cytotoxic alterations, upregulated the immunoexpression of PCNA and Ki-67, and restored the perturbed morphology of the hepatic and renal tissues. In conclusion, the obtained data demonstrated that SP co-administration elicits both protective and therapeutic potential against the FU-induced hepato- and nephropathy.
Sigma-Aldrich
Furan, ≥99%