Merck
  • Cobalt protoporphyrin IX increases endogenous G-CSF and mobilizes HSC and granulocytes to the blood.
Cobalt protoporphyrin IX increases endogenous G-CSF and mobilizes HSC and granulocytes to the blood.
EMBO molecular medicine (2019-11-12)
Agata Szade, Krzysztof Szade, Witold N Nowak, Karolina Bukowska-Strakova, Lucie Muchova, Monika Gońka, Monika Żukowska, Maciej Cieśla, Neli Kachamakova-Trojanowska, Marzena Rams-Baron, Alicja Ratuszna, Józef Dulak, Alicja Józkowicz
ABSTRACT
Granulocyte colony-stimulating factor (G-CSF) is used in clinical practice to mobilize cells from the bone marrow to the blood; however, it is not always effective. We show that cobalt protoporphyrin IX (CoPP) increases plasma concentrations of G-CSF, IL-6, and MCP-1 in mice, triggering the mobilization of granulocytes and hematopoietic stem and progenitor cells (HSPC). Compared with recombinant G-CSF, CoPP mobilizes higher number of HSPC and mature granulocytes. In contrast to G-CSF, CoPP does not increase the number of circulating T cells. Transplantation of CoPP-mobilized peripheral blood mononuclear cells (PBMC) results in higher chimerism and faster hematopoietic reconstitution than transplantation of PBMC mobilized by G-CSF. Although CoPP is used to activate Nrf2/HO-1 axis, the observed effects are Nrf2/HO-1 independent. Concluding, CoPP increases expression of mobilization-related cytokines and has superior mobilizing efficiency compared with recombinant G-CSF. This observation could lead to the development of new strategies for the treatment of neutropenia and HSPC transplantation.
Sigma-Aldrich
Protoporphyrin IX, ≥95%