Merck
  • In vitro biosynthesis of the Pseudomonas aeruginosa quorum-sensing signal molecule N-butanoyl-L-homoserine lactone.
In vitro biosynthesis of the Pseudomonas aeruginosa quorum-sensing signal molecule N-butanoyl-L-homoserine lactone.
Molecular microbiology (1998-05-21)
Y Jiang, M Camara, S R Chhabra, K R Hardie, B W Bycroft, A Lazdunski, G P Salmond, G S Stewart, P Williams
ABSTRACT
In Pseudomonas aeruginosa, synthesis of the quorum-sensing signal molecules N-butanoyl-L-homoserine lactone (BHL) and N-hexanoyl-L-homoserine lactone (HHL) requires the Luxl homologue Rhll(Vsml). By using thin-layer chromatography in conjunction with high-performance liquid chromatography (HPLC) and mass spectrometry, we show that purified Rhll can catalyse the biosynthesis of BHL and HHL using either S-adenosylmethionine (SAM) or homoserine lactone (HSL) but not homoserine as the source of the homoserine lactone moiety. As we were unable to detect homoserine lactone in cytoplasmic extracts of Escherichia coli, we conclude that SAM is the natural substrate for Rhll-directed N-acylhomoserine lactone (AHL) biosynthesis. The N-acyl chain of BHL and HHL can be supplied by the appropriately charged coenzyme A derivative (either n-butanoyl-CoA or n-hexanoyl-CoA). The specificity of Rhll for charged CoA derivatives is demonstrated as Rhll was unable to generate AHLs detectable in our bioassays from acetyl-CoA, malonyl-CoA, n-octanoyl-CoA, n-decanoyl-CoA, DL-beta-hydroxybutanoyl-CoA or crotonoyl-CoA. Rhll was also unable to use N-acetyl-S-3-oxobutanoylcysteamine, a chemical mimic for 3-oxobutanoyl-CoA. Furthermore, the Rhll-catalysed synthesis of BHL and HHL was most efficiently driven when NADPH was included in the reaction mixture.
Sigma-Aldrich
C4-HSL, ≥95% (HPLC)
Avanti
04:0 Coenzyme A, Avanti Research - A Croda Brand
Avanti
06:0 Coenzyme A, Avanti Research - A Croda Brand 870706P, powder