Merck
  • Structural basis of phosphatidylcholine recognition by the C2-domain of cytosolic phospholipase A2α.
Structural basis of phosphatidylcholine recognition by the C2-domain of cytosolic phospholipase A2α.
eLife (2019-05-03)
Yoshinori Hirano, Yong-Guang Gao, Daniel J Stephenson, Ngoc T Vu, Lucy Malinina, Dhirendra K Simanshu, Charles E Chalfant, Dinshaw J Patel, Rhoderick E Brown
ABSTRACT
Ca2+-stimulated translocation of cytosolic phospholipase A2α (cPLA2α) to the Golgi induces arachidonic acid production, the rate-limiting step in pro-inflammatory eicosanoid synthesis. Structural insights into the cPLA2α preference for phosphatidylcholine (PC)-enriched membranes have remained elusive. Here, we report the structure of the cPLA2α C2-domain (at 2.2 Å resolution), which contains bound 1,2-dihexanoyl-sn-glycero-3-phosphocholine (DHPC) and Ca2+ ions. Two Ca2+ are complexed at previously reported locations in the lipid-free C2-domain. One of these Ca2+ions, along with a third Ca2+, bridges the C2-domain to the DHPC phosphate group, which also interacts with Asn65. Tyr96 plays a key role in lipid headgroup recognition via cation-π interaction with the PC trimethylammonium group. Mutagenesis analyses confirm that Tyr96 and Asn65 function in PC binding selectivity by the C2-domain and in the regulation of cPLA2α activity. The DHPC-binding mode of the cPLA2α C2-domain, which differs from phosphatidylserine or phosphatidylinositol 4,5-bisphosphate binding by other C2-domains, expands and deepens knowledge of the lipid-binding mechanisms mediated by C2-domains.
Product Description
Avanti
18:1 SM (d18:1/18:1(9Z)), Avanti Research - A Croda Brand