Merck
  • Jadomycin B, an Aurora-B kinase inhibitor discovered through virtual screening.
Jadomycin B, an Aurora-B kinase inhibitor discovered through virtual screening.
Molecular cancer therapeutics (2008-08-30)
Da-Hua Fu, Wei Jiang, Jian-Ting Zheng, Gui-Yu Zhao, Yan Li, Hong Yi, Zhuo-Rong Li, Jian-Dong Jiang, Ke-Qian Yang, Yanchang Wang, Shu-Yi Si
ABSTRACT
Aurora kinases have emerged as promising targets for cancer therapy because of their critical role in mitosis. These kinases are well-conserved in all eukaryotes, and IPL1 gene encodes the single Aurora kinase in budding yeast. In a virtual screening attempt, 22 compounds were identified from nearly 15,000 microbial natural products as potential small-molecular inhibitors of human Aurora-B kinase. One compound, Jadomycin B, inhibits the growth of ipl1-321 temperature-sensitive mutant more dramatically than wild-type yeast cells, raising the possibility that this compound is an Aurora kinase inhibitor. Further in vitro biochemical assay using purified recombinant human Aurora-B kinase shows that Jadomycin B inhibits Aurora-B activity in a dose-dependent manner. Our results also indicate that Jadomycin B competes with ATP for the kinase domain, which is consistent with our docking prediction. Like other Aurora kinase inhibitors, Jadomycin B blocks the phosphorylation of histone H3 on Ser10 in vivo. We also present evidence suggesting that Jadomycin B induces apoptosis in tumor cells without obvious effects on cell cycle. All the results indicate that Jadomycin B is a new Aurora-B kinase inhibitor worthy of further investigation.
Sigma-Aldrich
Jadomycin B, ≥98% (HPLC)