Merck
  • A reversible, switchable pH-driven quaternary ammonium pillar[5]arene nanogate for mesoporous silica nanoparticles.
A reversible, switchable pH-driven quaternary ammonium pillar[5]arene nanogate for mesoporous silica nanoparticles.
Journal of materials chemistry. B (2019-12-24)
Evelyn C S Santos, Thiago C Dos Santos, Tamires S Fernandes, Fernanda L Jorge, Vanessa Nascimento, Vinicius G C Madriaga, Pâmella S Cordeiro, Noemi R Checca, Nathalia Meireles Da Costa, Luís Felipe Ribeiro Pinto, Célia M Ronconi
ABSTRACT
Here we describe the assembly and pH-driven operation of two nanocarriers based on non-functionalized (MCM-41) and carboxylate-functionalized (MCM-41-COOH) containers loaded with the anticancer drug doxorubicin (DOX) and capped by quaternary ammonium pillar[5]arene (P[5]A) nanogates. MCM-41 and MCM-41-COOH containers were synthesized and transmission and scanning electron microscopies showed nanoparticles with spherical morphology and dimensions of 85 ± 13 nm. The nanochannels of MCM-41 loaded with DOX were gated through the electrostatic interactions between P[5]A and the silanolate groups formed at the silica-water interface, yielding the MCM-41-DOX-P[5]A nanocarrier. The second nanocarrier was gated through the electrostatic interactions between the carboxylate groups mounted on the surface of MCM-41 and P[5]A, resulting in the MCM-41-COO-DOX-P[5]A nanocarrier. The DOX release profiles from both nanocarriers were investigated by UV-vis spectroscopy at different pH values (2.0, 5.5 and 7.4) and also in the presence of ions, such as citrate3- (19 mmol L-1) and Zn2+ (1.2 and 50 mmol L-1) at 37 °C. MCM-41-COO-DOX-P[5]A can be turned on and off eight times through the formation and breaking of electrostatic interactions. In vitro studies show that MCM-41-COO-DOX-P[5]A can penetrate and release DOX in the nucleus of human breast adenocarcinoma MCF-7 cancer cells leading to a pronounced cytotoxic effect. Therefore, the fabricated nanocarrier based on a water-soluble cationic pillar[5]arene nanogate, which is reversibly opened and closed by electrostatic interactions, can be considered as a promising drug transport and delivery technique for future cancer therapy.
Sigma-Aldrich
Yttrium aluminum oxide, nanopowder, <150 nm particle size (TEM), 99% trace metals basis
Sigma-Aldrich
3-Cyanopropyltriethoxysilane, 98%
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%