Merck
  • Molecular autopsy and family screening in a young case of sudden cardiac death reveals an unusually severe case of FHL1 related hypertrophic cardiomyopathy.
Molecular autopsy and family screening in a young case of sudden cardiac death reveals an unusually severe case of FHL1 related hypertrophic cardiomyopathy.
Molecular genetics & genomic medicine (2019-07-12)
Anna Gaertner-Rommel, Jens Tiesmeier, Thomas Jakob, Bernd Strickmann, Gunter Veit, Bernd Bachmann-Mennenga, Lech Paluszkiewicz, Karin Klingel, Uwe Schulz, Kai T Laser, Bernd Karger, Heidi Pfeiffer, Hendrik Milting
ABSTRACT
Hypertrophic cardiomyopathy (HCM) is a genetic cardiomyopathy with a prevalence of about 1:200. It is characterized by left ventricular hypertrophy, diastolic dysfunction and interstitial fibrosis; HCM might lead to sudden cardiac death (SCD) especially in the young. Due to low autopsy frequencies of sudden unexplained deaths (SUD) the true prevalence of SCD and especially of HCM among SUD remains unclear. Even in cases of proven SCD genetic testing is not a routine procedure precluding appropriate risk stratification and counseling of relatives. Here we report a case of SCD in a 19-year-old investigated by combined forensic and molecular autopsy. During autopsy of the index-patient HCM was detected. As no other possible cause of death could be uncovered by forensic autopsy the event was classified as SCD. Molecular autopsy identified two (probably) pathogenic genetic variants in FHL1 and MYBPC3. The MYBPC3 variant had an incomplete penetrance. The FHL1 variant was a de novo mutation. We detected reduced FHL1 mRNA levels and no FHL1 protein in muscle samples suggesting nonsense-mediated mRNA decay and/or degradation of the truncated protein in the SCD victim revealing a plausible disease mechanism. The identification of the genetic cause of the SCD contributed to the rational counseling of the relatives and risk assessment within the family. Furthermore our study revealed evidences for the pathomechanism of FHL1 mutations.
Sigma-Aldrich
Anti-FHL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1