Merck
  • Abnormal glycosylation of EAAT1 and EAAT2 in prefrontal cortex of elderly patients with schizophrenia.
Abnormal glycosylation of EAAT1 and EAAT2 in prefrontal cortex of elderly patients with schizophrenia.
Schizophrenia research (2009-09-01)
Deborah Bauer, Vahram Haroutunian, James H Meador-Woodruff, Robert E McCullumsmith
ABSTRACT
The excitatory amino acid transporters (EAATs) are a family of molecules that are essential for regulation of synaptic glutamate levels. The EAATs may also be regulated by N-glycosylation, a posttranslational modification that is critical for many cellular functions including localization in the plasma membrane. We hypothesized that glycosylation of the EAATs is abnormal in schizophrenia. To test this hypothesis, we treated postmortem tissue from the dorsolateral prefrontal and anterior cingulate cortices of patients with schizophrenia and comparison subjects with deglycosylating enzymes. We then measured the resulting shifts in molecular weight of the EAATs using Western blot analysis to determine the mass of glycans cleaved from the transporter. We found evidence for less glycosylation of both EAAT1 and EAAT2 in schizophrenia. We did not detect N-linked glycosylation of EAAT3 in either schizophrenia or the comparison subjects in these regions. Our data suggest an abnormality of posttranslational modification of glutamate transporters in schizophrenia that suggests a decreased capacity for glutamate reuptake.
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor m4 Antibody, clone 18C7.2, clone 18C7.2, Chemicon®, from mouse
Pricing and availability is not currently available.