Merck
  • ROBO2 is a stroma suppressor gene in the pancreas and acts via TGF-β signalling.
ROBO2 is a stroma suppressor gene in the pancreas and acts via TGF-β signalling.
Nature communications (2018-12-07)
Andreia V Pinho, Mathias Van Bulck, Lorraine Chantrill, Mehreen Arshi, Tatyana Sklyarova, David Herrmann, Claire Vennin, David Gallego-Ortega, Amanda Mawson, Marc Giry-Laterriere, Astrid Magenau, Gunther Leuckx, Luc Baeyens, Anthony J Gill, Phoebe Phillips, Paul Timpson, Andrew V Biankin, Jianmin Wu, Ilse Rooman
ABSTRACT
Whereas genomic aberrations in the SLIT-ROBO pathway are frequent in pancreatic ductal adenocarcinoma (PDAC), their function in the pancreas is unclear. Here we report that in pancreatitis and PDAC mouse models, epithelial Robo2 expression is lost while Robo1 expression becomes most prominent in the stroma. Cell cultures of mice with loss of epithelial Robo2 (Pdx1Cre;Robo2F/F) show increased activation of Robo1+ myofibroblasts and induction of TGF-β and Wnt pathways. During pancreatitis, Pdx1Cre;Robo2F/F mice present enhanced myofibroblast activation, collagen crosslinking, T-cell infiltration and tumorigenic immune markers. The TGF-β inhibitor galunisertib suppresses these effects. In PDAC patients, ROBO2 expression is overall low while ROBO1 is variably expressed in epithelium and high in stroma. ROBO2low;ROBO1high patients present the poorest survival. In conclusion, Robo2 acts non-autonomously as a stroma suppressor gene by restraining myofibroblast activation and T-cell infiltration. ROBO1/2 expression in PDAC patients may guide therapy with TGF-β inhibitors or other stroma /immune modulating agents.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Vimentin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture