Merck
  • Redirected T-cell killing of solid cancers targeted with an anti-CD3/Trop-2-bispecific antibody is enhanced in combination with interferon-α.
Redirected T-cell killing of solid cancers targeted with an anti-CD3/Trop-2-bispecific antibody is enhanced in combination with interferon-α.
Molecular cancer therapeutics (2014-07-24)
Edmund A Rossi, Diane L Rossi, Thomas M Cardillo, Chien-Hsing Chang, David M Goldenberg
ABSTRACT
Trop-2 has limited presence on normal tissues but is highly expressed in diverse epithelial cancers. (E1)-3s is a T-cell-redirecting trivalent bispecific antibody (bsAb), comprising an anti-CD3 scFv covalently linked to a stabilized dimer of a Trop-2-targeting Fab using Dock-and-Lock. We show for the first time that bsAb-mediated bidirectional trogocytosis occurs between target and T cells and involves immunologic synapses. We studied the effects of interferon-α (INFα) on (E1)-3s-mediated T-cell killing of human gastric and pancreatic cancer cell lines. T-cell activation, cytokine induction, and cytotoxicity were evaluated ex vivo using peripheral blood mononuclear cells (PBMC) or T cells with NCI-N87 gastric cancer as target cells. In vivo activity was assayed with NCI-N87 and Capan-1 (pancreatic) xenografts. In the presence of target cells and PBMCs, (E1)-3s did not cause excess cytokine production. When combined with (E1)-3s, peginterferonalfa-2a--which alone did not increase T-cell activation or raise cytokine levels over baseline--increased CD69 expression but did not significantly increase cytokine induction. (E1) 3s mediated a highly potent T-cell lysis of NCI-N87 target cells in vitro. Inclusion of peginterferonalfa-2a or a more potent form of INFα, 20*-2b, significantly potentiated the activity of (E1)-3s by more than 2.5- or 7-fold, respectively. In vivo, combining peginterferonalfa-2a with (E1)-3s delayed Capan-1 growth longer than each single agent. Similarly, combination therapy delayed tumor proliferation of NCI-N87 compared with (E1)-3s or peginterferonalfa-2a single-treatment groups. (E1)-3s effectively induced T-cell-mediated killing of Trop-2-expressing pancreatic and gastric cancers, which was enhanced with INFα.