Merck
  • Leukemia growth is inhibited by benzoxime without causing any harmful effect in rats bearing RBL-1 ×enotransplants.
Leukemia growth is inhibited by benzoxime without causing any harmful effect in rats bearing RBL-1 ×enotransplants.
Oncology letters (2019-01-25)
Yingchun Li, Huihan Wang, Rong Zhang, Guojun Zhang, Ying Yang, Zhuogang Liu
ABSTRACT
The present study aimed to investigate the effect of benzoxime on leukemia RBL-1 cell proliferation and a leukemic Sprague-Dawley rat model. Proliferation of RBL-1 cells was determined using an MTT assay. Sprague-Dawley rats were assigned randomly into three groups of 10 animals each, where the positive control group was administered an intravenous injection of normal saline, the negative control group was administered 1×106 RBL-1 cells and the treatment group was administered with 1×106 RBL-1 cells and then benzoxime (50 mg/kg/day) for 1 week. Increased dosage of benzoxime reduced RBL-1 cell viability from 92 at 2 µM to ٢١٪ at ١٢ µM after ٢٤ h. Benzoxime treatment prevented the loss of body weight in the rats with leukemia. Compared with the negative control rats, the body weight was determined to be significantly reduced (P<0.05) in the positive control rats. The weight of the spleen and liver was determined to be significantly increased (P<0.02) in the positive control rats and the benzoxime-treated rats compared with that in the negative control group on day 35 of RBL-1 cell implantation. Analysis of leukocytes in rats on day 35 demonstrated a significant reduction (P<0.05) in the cluster of differentiation (CD)11b and CD45 level in the positive control group compared with that in the negative control group. The level of CD11b and CD45 was determined to be similar in the rats in the benzoxime treatment and negative control groups. Analysis of the level of serum glutamic pyruvic transaminase, serum glutamic oxaloacetic transaminase and blood urea nitrogen indicated that all three components exhibited no significant changes in the rats following treatment with benzoxime compared with the component levels in the negative control group. The levels of these three components were in the normal range in rats treated with benzoxime on day 35 of cell implantation. These data demonstrated that the liver and kidneys are not influenced by benzoxime in rats with leukemia. In summary, the present study demonstrated that benzoxime efficiently prevents leukemia growth without inducing any harmful effects in rat models through targeting CD11b and CD45 level; thus, benzoxime should be evaluated further regarding its use in the treatment of leukemia.
Sigma-Aldrich
Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-CD19 antibody, Rabbit monoclonal, clone SP110, recombinant, expressed in proprietary host, affinity isolated antibody