- A Sendai virus recombinant vaccine expressing a gene for truncated human metapneumovirus (hMPV) fusion protein protects cotton rats from hMPV challenge.
A Sendai virus recombinant vaccine expressing a gene for truncated human metapneumovirus (hMPV) fusion protein protects cotton rats from hMPV challenge.
Virology (2017-06-13)
Charles J Russell, Bart G Jones, Robert E Sealy, Sherri L Surman, John N Mason, Randall T Hayden, Ralph A Tripp, Toru Takimoto, Julia L Hurwitz
PMID28605636
ABSTRACT
Human metapneumovirus (hMPV) infections pose a serious health risk to young children, particularly in cases of premature birth. No licensed vaccine exists and there is no standard treatment for hMPV infections apart from supportive hospital care. We describe the production of a Sendai virus (SeV) recombinant that carries a gene for a truncated hMPV fusion (F) protein (SeV-MPV-Ft). The vaccine induces binding and neutralizing antibody responses toward hMPV and protection against challenge with hMPV in a cotton rat system. Results encourage advanced development of SeV-MPV-Ft to prevent the morbidity and mortality caused by hMPV infections in young children.
Sigma-Aldrich
Anti-Human Metapneumovirus Antibody, clone 132, clone 132, Chemicon®, from mouse