- In vitro and in vivo inhibitory actions of morin on rat brain phosphatidylinositolphosphate kinase activity.
In vitro and in vivo inhibitory actions of morin on rat brain phosphatidylinositolphosphate kinase activity.
Phosphatidylinositol-4,5-bisphosphate occupies a central role in signal transduction and in cellular transformation. Phosphatidylinositol-4,5-bisphosphate is produced by the enzymatic phosphorylation of phosphatidylinositol-4-phosphate by phosphatidylinositolphosphate kinase (EC 2.7.1.68). Inhibition of this enzyme might conceivably lowers the cellular pool of phosphatidylinositol-4,5-bisphosphate, thus constituting a feasible control point in regulating signal transduction and cellular transformation. Morin, a plant flavonoid, was demonstrated to exhibit in vitro inhibitory action on phosphatidylinositolphosphate kinase extracted from rat brain. This inhibition of enzymatic activity was found to be dose-dependent, with an IC50 value of approximately 10 microM morin. Lineweaver-Burk transformation of the inhibition data indicates that inhibition was competitive with respect to ATP. The Ki was calculated to be 5.15 x 10(-6) M. Inhibition was uncompetitive with respect to phosphatidylinositol-4-phosphate. The Ki was determined to be 0.94 x 10(-5) M. Administration of morin to rats led to a decrease in phosphatidylinositolphosphate kinase activity in brain extracts. This in vivo action of morin was found to be dose-dependent and time-dependent. These effects of morin on rat brain phosphatidylinositolphosphate kinase activity are discussed in relation to the other reported biological actions of this flavonoid.