Merck
  • Treadmill Exercise Decreases Aβ Deposition and Counteracts Cognitive Decline in APP/PS1 Mice, Possibly via Hippocampal Microglia Modifications.
Treadmill Exercise Decreases Aβ Deposition and Counteracts Cognitive Decline in APP/PS1 Mice, Possibly via Hippocampal Microglia Modifications.
Frontiers in aging neuroscience (2019-04-27)
Xianliang Zhang, Qiang He, Tao Huang, Na Zhao, Fei Liang, Bo Xu, Xianghe Chen, Tuojian Li, Jianzhong Bi
ABSTRACT
Recent studies have suggested that exercise may be beneficial for delaying or attenuating Alzheimer's disease (AD). However, the underlying mechanisms were not clear. Microglia-mediated neuroinflammation is suggested to play an important role in the pathology of AD. The present study investigated the beneficial effects of treadmill exercise on amyloid-β (Aβ) deposition and cognitive function in amyloid precursor protein (APP)/PS1 mice in the early stage of AD progression and microglia-mediated neuroinflammation was mainly analyzed. The results demonstrated that 12 weeks of treadmill exercise preserved hippocampal cognitive function in APP/PS1 mice and substantially suppressed Aβ accumulation in the hippocampus. Treadmill exercise significantly inhibited neuroinflammation, which was characterized by a remarkably reduced expression of pro-inflammatory factors and increased expression of anti-inflammatory mediators in the hippocampus, resulting from a shift in activated microglia from the M1 to M2 phenotype. Treadmill exercise also attenuated oxidative stress presented by a marked reduction in methane dicarboxylic aldehyde (MDA) level and dramatically elevated SOD and Mn-SOD activities in the hippocampus. These findings suggest that treadmill exercise can effectively prevent the decrease in hippocampal-dependent cognitive function and Aβ deposits in early AD progression possibly via modulating microglia-mediated neuroinflammation and oxidative stress.
Sigma-Aldrich
Monoclonal Anti-Iba1 antibody produced in mouse, clone GT10312, affinity isolated antibody
Sigma-Aldrich
Anti-Amyloid β Antibody, clone W0-2, clone WO2, from mouse