Merck
  • Dynamic Effect of Fresh and Aged Biochar on the Behavior of the Herbicide Mesotrione in Soils.
Dynamic Effect of Fresh and Aged Biochar on the Behavior of the Herbicide Mesotrione in Soils.
Journal of agricultural and food chemistry (2019-08-06)
Beatriz Gámiz, Pilar Velarde, Kurt A Spokas, Lucía Cox
ABSTRACT
In this study, we assessed the sorption, dissipation, and leaching of the herbicide mesotrione in soil amended with fresh and field-aged biochars, when added to the soil. The aging process was performed by burying the fresh biochar at 10 cm depth in three soils located in different points across the USA [Wisconsin (ABC_WI), Idaho (ABC_ID), and South Carolina (ABC_SC)] for six months. ABC_ID and ABC_SC slightly increased the sorption of mesotrione in soils, whereas ABC_WI removed greater amounts of herbicide from the solution. This was attributed to differences in water-soluble components and metal content of this aged biochar. Consequently, the persistence of the herbicide in the amended soils with fresh biochar and ABC_ID and ABC_SC were similar to that in unamended soils, while ABC_WI slightly increased mesotrione half-life. Differences between treatments were detected in leaching studies although no direct relationship with the dissipation batch studies was observed. Mesotrione leaching could not be detected in soil columns amended with ABC_WI and was high for the rest of treatments. The outcomes from this work demonstrate that temporal variability of biochar sorption capacities due to soil exposure can occur altering mesotrione's behavior in biochar-amended soils.
Sigma-Aldrich
2-Amino-4-(methylsulfonyl)benzoic acid, AldrichCPR
Supelco
Mesotrione, PESTANAL®, analytical standard