Merck
  • Decreased plasma phospholipid concentrations and increased acid sphingomyelinase activity are accurate biomarkers for community-acquired pneumonia.
Decreased plasma phospholipid concentrations and increased acid sphingomyelinase activity are accurate biomarkers for community-acquired pneumonia.
Journal of translational medicine (2019-11-13)
Haroon Arshad, Juan Carlos López Alfonso, Raimo Franke, Katina Michaelis, Leonardo Araujo, Aamna Habib, Yuliya Zboromyrska, Eva Lücke, Emilia Strungaru, Manas K Akmatov, Haralampos Hatzikirou, Michael Meyer-Hermann, Astrid Petersmann, Matthias Nauck, Mark Brönstrup, Ursula Bilitewski, Laurent Abel, Jorg Sievers, Jordi Vila, Thomas Illig, Jens Schreiber, Frank Pessler
ABSTRACT
There continues to be a great need for better biomarkers and host-directed treatment targets for community-acquired pneumonia (CAP). Alterations in phospholipid metabolism may constitute a source of small molecule biomarkers for acute infections including CAP. Evidence from animal models of pulmonary infections and sepsis suggests that inhibiting acid sphingomyelinase (which releases ceramides from sphingomyelins) may reduce end-organ damage. We measured concentrations of 105 phospholipids, 40 acylcarnitines, and 4 ceramides, as well as acid sphingomyelinase activity, in plasma from patients with CAP (n = 29, sampled on admission and 4 subsequent time points), chronic obstructive pulmonary disease exacerbation with infection (COPD, n = 13) as a clinically important disease control, and 33 age- and sex-matched controls. Phospholipid concentrations were greatly decreased in CAP and normalized along clinical improvement. Greatest changes were seen in phosphatidylcholines, followed by lysophosphatidylcholines, sphingomyelins and ceramides (three of which were upregulated), and were least in acylcarnitines. Changes in COPD were less pronounced, but also differed qualitatively, e.g. by increases in selected sphingomyelins. We identified highly accurate biomarkers for CAP (AUC ≤ 0.97) and COPD (AUC ≤ 0.93) vs. Controls, and moderately accurate biomarkers for CAP vs. COPD (AUC ≤ 0.83), all of which were phospholipids. Phosphatidylcholines, lysophosphatidylcholines, and sphingomyelins were also markedly decreased in S. aureus-infected human A549 and differentiated THP1 cells. Correlations with C-reactive protein and procalcitonin were predominantly negative but only of mild-to-moderate extent, suggesting that these markers reflect more than merely inflammation. Consistent with the increased ceramide concentrations, increased acid sphingomyelinase activity accurately distinguished CAP (fold change = 2.8, AUC = 0.94) and COPD (1.75, 0.88) from Controls and normalized with clinical resolution. The results underscore the high potential of plasma phospholipids as biomarkers for CAP, begin to reveal differences in lipid dysregulation between CAP and infection-associated COPD exacerbation, and suggest that the decreases in plasma concentrations are at least partially determined by changes in host target cells. Furthermore, they provide validation in clinical blood samples of acid sphingomyelinase as a potential treatment target to improve clinical outcome of CAP.
Product Description
Avanti
C24:1 Ceramide (d18:1/24:1(15Z)), Avanti Research - A Croda Brand
Avanti
C16 Ceramide (d18:1/16:0), Avanti Research - A Croda Brand
Avanti
C18 Ceramide (d18:1/18:0), Avanti Research - A Croda Brand
Avanti
C24 Ceramide (d18:1/24:0), Avanti Research - A Croda Brand