Merck
  • Identification of potent and selective inhibitors of PDGF receptor autophosphorylation.
Identification of potent and selective inhibitors of PDGF receptor autophosphorylation.
Journal of medicinal chemistry (2006-03-31)
Takayuki Furuta, Teruyuki Sakai, Terufumi Senga, Tatsushi Osawa, Kazuo Kubo, Toshiyuki Shimizu, Rika Suzuki, Tetsuya Yoshino, Megumi Endo, Atsushi Miwa
ABSTRACT
We report the structure-activity relationship of quinoline and quinazoline derivatives, which include urea, thiourea, urethane, and acylthiourea groups, as inhibitors of the platelet-derived growth factor (PDGF) receptor autophosphorylation. Our previous studies showed that the quinoline and quinazoline derivatives including urea, thiourea, and carbamate groups were highly potent compounds as the PDGF receptor autophosphorylation inhibitor, but these compounds did not exhibit receptor selectivity between the PDGF receptor and the c-kit receptor. As a result of further synthesis and biological evaluation, we have found that the quinoline and quinazoline-acylthiourea derivatives showed not only good inhibitory activity for the PDGF receptor but also receptor selectivity between the PDGF receptor and the c-kit receptor. Furthermore N-{4-[(6,7-dimethoxy-4-quinolyl)oxy]phenyl}-N'-(2-methylbenzoyl)thiourea exhibited potent oral efficacy in in vivo assay using the rat carotid balloon injury model. Therefore, the quinoline and quinazoline-acylthiourea derivatives may be expected to have potential as therapeutic agents for the treatment of restenosis.
Sigma-Aldrich
PDGFR Tyrosine Kinase Inhibitor V, The PDGFR Tyrosine Kinase Inhibitor V, also referenced under CAS 347155-76-4, controls the biological activity of PDGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Pricing and availability is not currently available.