Merck
  • Dafachronic acid promotes larval development in Haemonchus contortus by modulating dauer signalling and lipid metabolism.
Dafachronic acid promotes larval development in Haemonchus contortus by modulating dauer signalling and lipid metabolism.
PLoS pathogens (2019-07-25)
Guangxu Ma, Tao Wang, Pasi K Korhonen, Neil D Young, Shuai Nie, Ching-Seng Ang, Nicholas A Williamson, Gavin E Reid, Robin B Gasser
ABSTRACT
Here, we discovered an endogenous dafachronic acid (DA) in the socioeconomically important parasitic nematode Haemonchus contortus. We demonstrate that DA promotes larval exsheathment and development in this nematode via a relatively conserved nuclear hormone receptor (DAF-12). This stimulatory effect is dose- and time-dependent, and relates to a modulation of dauer-like signalling, and glycerolipid and glycerophospholipid metabolism, likely via a negative feedback loop. Specific chemical inhibition of DAF-9 (cytochrome P450) was shown to significantly reduce the amount of endogenous DA in H. contortus; compromise both larval exsheathment and development in vitro; and modulate lipid metabolism. Taken together, this evidence shows that DA plays a key functional role in the developmental transition from the free-living to the parasitic stage of H. contortus by modulating the dauer-like signalling pathway and lipid metabolism. Understanding the intricacies of the DA-DAF-12 system and associated networks in H. contortus and related parasitic nematodes could pave the way to new, nematode-specific treatments.