Merck
  • Semaphorin 4A acts in a feed-forward loop with NF-κB pathway to exacerbate catabolic effect of IL-1β on chondrocytes.
Semaphorin 4A acts in a feed-forward loop with NF-κB pathway to exacerbate catabolic effect of IL-1β on chondrocytes.
International immunopharmacology (2019-01-28)
Hua Zhang, Qiushi Wei, Xiaobing Xiang, Bengen Zhou, Jianfa Chen, Jie Li, Qihuo Li, Hao Xiong, Fuyifei Liu
ABSTRACT
Inflammation is fundamental in osteoarthritis (OA) pathogenesis. Semaphorin 4A (Sema4A) has been implicated in immune-associated diseases, however, its role in OA remains unclear. In this study, we show that Sema4A is upregulated in knee OA articular cartilage as well as in chondrocytes exposed to IL-1β treatment in vitro. Moreover, IL-1β-induced Sema4A upregulation is abrogated in the presence of BAY 11-7082, a specific inhibitor of NF-κB pathway, suggesting that the activation of NF-κB is required for Sema4A upregulation under this pathological condition. Intriguingly, Sema4A in turn activates NF-κB through facilitating Rac1/AKT-dependent IκBα phosphorylation and subsequent degradation. Functionally, Sema4A aggravates the catabolic effect of IL-1β on chondrocytes, which can be largely attributed to exacerbated NF-κB activation, since NF-κB inhibition remarkably abolishes this effect. In conclusion, our study suggests that Sema4A is a novel regulator of NF-κB-dependent catabolic events in chondrocytes, which may underlie OA pathogenesis.