Merck
  • Monoacylglycerol Lipase Inactivation by Using URB602 Mitigates Myocardial Damage in a Rat Model of Cardiac Arrest.
Monoacylglycerol Lipase Inactivation by Using URB602 Mitigates Myocardial Damage in a Rat Model of Cardiac Arrest.
Critical care medicine (2018-11-16)
Kerong Hai, Guo Chen, Xueyan Gou, Haixia Jang, Deying Gong, Yan Cheng, Chansheng Gong, Xinghuan Li, Yuqi Liu, Huan Li, Gang Zhang, Linghui Yang, Bowen Ke, Jin Liu
ABSTRACT
Monoacylglycerol lipase participates in organ protection by regulating the hydrolysis of the endocannabinoid 2-arachidonoylglycerol. This study investigated whether blocking monoacylglycerol lipase protects against postresuscitation myocardial injury and improves survival in a rat model of cardiac arrest and cardiopulmonary resuscitation. Prospective randomized laboratory study. University research laboratory. Male Sprague-Dawley rat (n = 96). Rats underwent 8-minute asphyxia-based cardiac arrest and resuscitation. Surviving rats were randomly divided into cardiopulmonary resuscitation + URB602 group, cardiopulmonary resuscitation group, and sham group. One minute after successful resuscitation, rats in the cardiopulmonary resuscitation + URB602 group received a single dose of URB602 (5 mg/kg), a small-molecule monoacylglycerol lipase inhibitor, whereas rats in the cardiopulmonary resuscitation group received an equivalent volume of vehicle solution. The sham rats underwent all of the procedures performed on rats in the cardiopulmonary resuscitation and cardiopulmonary resuscitation + URB602 groups minus cardiac arrest and asphyxia. Survival was recorded 168 hours after the return of spontaneous circulation (n = 22 in each group). Compared with vehicle treatment (31.8%), URB602 treatment markedly improved survival (63.6%) 168 hours after cardiopulmonary resuscitation. Next, we used additional surviving rats to evaluate myocardial and mitochondrial injury 6 hours after return of spontaneous circulation, and we found that URB602 significantly reduced myocardial injury and prevented myocardial mitochondrial damage. In addition, URB602 attenuated the dysregulation of endocannabinoid and eicosanoid metabolism 6 hours after return of spontaneous circulation and prevented the acceleration of mitochondrial permeability transition 15 minutes after return of spontaneous circulation. Monoacylglycerol lipase blockade may reduce myocardial and mitochondrial injury and significantly improve the resuscitation effect after cardiac arrest and cardiopulmonary resuscitation.