Merck
  • Fractional carbon dioxide laser assisted delivery of topical tazarotene versus topical tioconazole in the treatment of onychomycosis.
Fractional carbon dioxide laser assisted delivery of topical tazarotene versus topical tioconazole in the treatment of onychomycosis.
The Journal of dermatological treatment (2018-08-08)
Essam Bakr Abd El-Aal, Hamed Mohamed Abdo, Shady Mahmoud Ibrahim, Mostafa Taha Eldestawy
ABSTRACT
Onychomycosis is a chronic fungal infection of the nails, and the treatment has been proven to be a challenge to healthcare professionals. To evaluate the efficacy of fractional carbon dioxide laser-assisted delivery of topical tazarotene versus topical tioconazole in the treatment of onychomycosis. A total of 102 patients with onychomycosis were randomly assigned to groups A and B, and both groups were treated with four sessions of fractional CO2 laser and followed by topical tazarotene 0.1% in Group A and topical tioconazole 28% in Group B. The clinical effect, KOH examination, and culture for the affected nails in the two groups were analyzed. One month after the last session, regarding clinical response, 35.3% showed complete improvement in Group A versus 33.3% in Group B without significant difference. There was a significant difference between the two studied groups as regards KOH test and culture result before and after treatment (p value<.001), they were turned negative in 91.7% and 100% of patients in Group A and 78.3% and 95.5% of patients in Group B, respectively. Fractional CO2 laser-assisted drug delivery is an effective and safe treatment option for onychomycosis.