Merck
  • SOX2-Dependent Transcription in Clock Neurons Promotes the Robustness of the Central Circadian Pacemaker.
SOX2-Dependent Transcription in Clock Neurons Promotes the Robustness of the Central Circadian Pacemaker.
Cell reports (2019-03-21)
Arthur H Cheng, Pascale Bouchard-Cannon, Sara Hegazi, Christopher Lowden, Samuel W Fung, Cheng-Kang Chiang, Rob W Ness, Hai-Ying Mary Cheng
ABSTRACT
Clock neurons within the mammalian suprachiasmatic nuclei (SCN) encode circadian time using interlocked transcription-translation feedback loops (TTFLs) that drive rhythmic gene expression. However, the contributions of other transcription factors outside of the circadian TTFLs to the functionality of the SCN remain obscure. Here, we report that the stem and progenitor cell transcription factor, sex-determining region Y-box 2 (SOX2), is expressed in adult SCN neurons and positively regulates transcription of the core clock gene, Period2. Mice lacking SOX2 selectively in SCN neurons display imprecise, poorly consolidated behavioral rhythms that do not entrain efficiently to environmental light cycles and that are highly susceptible to constant light-induced arrhythmicity. RNA sequencing revealed that Sox2 deficiency alters the SCN transcriptome, reducing the expression of core clock genes and neuropeptide-receptor systems. By defining the transcriptional landscape within SCN neurons, SOX2 enables the generation of robust, entrainable circadian rhythms that accurately reflect environmental time.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Vasopressin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-MeCP2 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-CLOCK Antibody, serum, Chemicon®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution