- Occurrence of fatty acid epoxide hydrolases in soybean (Glycine max). Purification and characterization of the soluble form.
Occurrence of fatty acid epoxide hydrolases in soybean (Glycine max). Purification and characterization of the soluble form.
The Biochemical journal (1992-03-15)
E Blée, F Schuber
PMID1554352
ABSTRACT
Epoxide hydrolases catalysing the hydration of cis-9,10-epoxystearate into threo-9,10-dihydroxystearate have been detected in soybean (Glycine max) seedlings. The major activity was found in the cytosol, a minor fraction being strongly associated with microsomes. The soluble enzyme, which was purified to apparent homogeneity by (NH4)2SO4 fractionation, hydrophobic, DEAE- and gel-filtration chromatographies, has a molecular mass of 64 kDa and a pI of 5.4.
MATERIALS