Merck
  • Congenital jaundice in rats with a mutation in a multidrug resistance-associated protein gene.
Congenital jaundice in rats with a mutation in a multidrug resistance-associated protein gene.
Science (New York, N.Y.) (1996-02-23)
C C Paulusma, P J Bosma, G J Zaman, C T Bakker, M Otter, G L Scheffer, R J Scheper, P Borst, R P Oude Elferink
ABSTRACT
The human Dubin-Johnson syndrome and its animal model, the TR(-) rat, are characterized by a chronic conjugated hyperbilirubinemia. TR(-) rats are defective in the canalicular multispecific organic anion transporter (cMOAT), which mediates hepatobiliary excretion of numerous organic anions. The complementary DNA for rat cmoat, a homolog of the human multidrug resistance gene (hMRP1), was isolated and shown to be expressed in the canalicular membrane of hepatocytes. In the TR(-) rat, a single-nucleotide deletion in this gene resulted in a reduced messenger RNA level and absence of the protein. It is likely that this mutation accounts for the TR(-) phenotype.
Sigma-Aldrich
Anti-MRP2 Antibody, clone M2 I-4, culture supernatant, clone M2 I-4, Chemicon®
Sigma-Aldrich
Anti-MRP2 Antibody, CT, clone M2 III-6, culture supernatant, clone M2 III-6, Chemicon®