Merck
  • Interleukin-1 in lipopolysaccharide induced chorioamnionitis in the fetal sheep.
Interleukin-1 in lipopolysaccharide induced chorioamnionitis in the fetal sheep.
Reproductive sciences (Thousand Oaks, Calif.) (2011-04-16)
Clare A Berry, Ilias Nitsos, Noah H Hillman, J Jane Pillow, Graeme R Polglase, Boris W Kramer, Matthew W Kemp, John P Newnham, Alan H Jobe, Suhas G Kallapur
ABSTRACT
We tested the hypothesis that interleukin 1 (IL-1) mediates intra-amniotic lipopolysaccharide (LPS)-induced chorioamnionitis in preterm fetal sheep. Time-mated Merino ewes with singleton fetuses received IL-1α, LPS, or saline (control) by intra-amniotic injection 1 to 2 days before operative delivery at 124 ± 1 days gestational age (N = 5-9/group; term = 150 days). Recombinant human IL-1 receptor antagonist (rhIL-1ra) was given into the amniotic fluid 3 hours before intra-amniotic LPS or saline to block IL-1 signaling. Inflammation in the chorioamnion was determined by histology, cytokine messenger RNA (mRNA), protein expression, and by quantitation of activated inflammatory cells. Intra-amniotic IL-1 and LPS both induced chorioamnionitis. However, IL-1 blockade with IL-1ra did not decrease intra-amniotic LPS-induced increases in pro-inflammatory cytokine mRNAs, numbers of inflammatory cells, myeloperoxidase, or monocyte chemotactic protein-1-expressing cells in the chorioamnion. We conclude that IL-1 and LPS both can cause chorioamnionitis, but IL-1 is not an important mediator of LPS-induced chorioamnionitis in fetal sheep.
Sigma-Aldrich
Anti-Interleukin-6 Antibody, serum, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Interleukin-8 Antibody, serum, Chemicon®
Pricing and availability is not currently available.