Merck
  • Increased proteolytic resistance of ribonuclease A by protein engineering.
Increased proteolytic resistance of ribonuclease A by protein engineering.
Protein engineering (2001-12-12)
Y Markert, J Köditz, J Mansfeld, U Arnold, R Ulbrich-Hofmann
ABSTRACT
Although highly stable toward unfolding, native ribonuclease A is known to be cleaved by unspecific proteases in the flexible loop region near Ala20. With the aim to create a protease-resistant ribonuclease A, Ala20 was substituted for Pro by site-directed mutagenesis. The resulting mutant enzyme was nearly identical to the wild-type enzyme in the near-UV and far-UV circular dichroism spectra, in its activity to 2',3'-cCMP and in its thermodynamic stability. However, the proteolytic resistance to proteinase K and subtilisin Carlsberg was extremely increased. Pseudo-first-order rate constants of proteolysis, determined by densitometric analysis of the bands of intact protein in SDS-PAGE, decreased by two orders of magnitude. In contrast, the rate constant of proteolysis with elastase was similar to that of the wild-type enzyme. These differences can be explained by the analysis of the fragments occurring in proteolysis with elastase. Ser21-Ser22 was identified as the main primary cleavage site in the degradation of the mutant enzyme by elastase. Obviously, this bond is not cleavable by proteinase K or subtilisin Carlsberg. The results demonstrate the high potential of a single mutation in protein stabilization to proteolytic degradation.
Sigma-Aldrich
Cytidine 2′:3′-cyclic monophosphate monosodium salt, ≥95% (HPLC)