Merck
  • Design and synthesis of novel dual-action compounds targeting the adenosine A(2A) receptor and adenosine transporter for neuroprotection.
Design and synthesis of novel dual-action compounds targeting the adenosine A(2A) receptor and adenosine transporter for neuroprotection.
ChemMedChem (2011-06-22)
Jhih-Bin Chen, Eric Minwei Liu, Ting-Rong Chern, Chieh-Wen Yang, Chia-I Lin, Nai-Kuei Huang, Yun-Lian Lin, Yijuang Chern, Jung-Hsin Lin, Jim-Min Fang
ABSTRACT
A novel compound, N⁶-(4-hydroxybenzyl)adenosine, isolated from Gastrodia elata and which has been shown to be a potential therapeutic agent for preventing and treating neurodegenerative disease, was found to target both the adenosine A(2A) receptor (A(2A) R) and the equilibrative nucleoside transporter 1 (ENT1). As A(2A) R and ENT1 are proximal in the synaptic crevice of striatum, where the mutant huntingtin aggregate is located, the dual-action compounds that concomitantly target these two membrane proteins may be beneficial for the therapy of Huntington's disease. To design the desired dual-action compounds, pharmacophore models of the A(2A) R agonists and the ENT1 inhibitors were constructed. Accordingly, potentially active compounds were designed and synthesized by chemical modification of adenosine, particularly at the N⁶ and C⁵' positions, if the predicted activity was within an acceptable range. Indeed, some of the designed compounds exhibit significant dual-action properties toward both A(2A) R and ENT1. Both pharmacophore models exhibit good statistical correlation between predicted and measured activities. In agreement with competitive ligand binding assay results, these compounds also prevent apoptosis in serum-deprived PC12 cells, rendering a crucial function in neuroprotection and potential utility in the treatment of neurodegenerative diseases.
Sigma-Aldrich
N6-(4-Hydroxybenzyl) adenine riboside, ≥98% (HPLC)
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis