Merck
  • DAST-mediated cyclization of α,α-disubstituted-α-acylaminoketones: efficient and divergent synthesis of unprecedented heterocycles.
DAST-mediated cyclization of α,α-disubstituted-α-acylaminoketones: efficient and divergent synthesis of unprecedented heterocycles.
Organic letters (2010-12-15)
Aurélien Bigot, Judith Blythe, Chirag Pandya, Trixie Wagner, Olivier Loiseleur
ABSTRACT
The design of a new potent nonsteroidal ecdysone agonist led to the discovery of a diethylaminosulfur trifluoride (DAST)-mediated cyclization of α,α-disubstituted-α-acylaminoketones. The resulting fluorooxazolines can be ring-opened or selectively substituted by a range of nucleophiles to provide in high yields a diverse array of unprecedented heterocyclic frameworks.
Sigma-Aldrich
(Diethylamino)sulfur trifluoride, 95%
Pricing and availability is not currently available.