Merck
  • Histone modifications in fatty acid synthase modulated by carbohydrate responsive element binding protein are associated with non‑alcoholic fatty liver disease.
Histone modifications in fatty acid synthase modulated by carbohydrate responsive element binding protein are associated with non‑alcoholic fatty liver disease.
International journal of molecular medicine (2018-05-23)
Can Cai, Huihong Yu, Guangming Huang, Xuan Du, Xiaoqing Yu, Youping Zhou, Wei Shen
ABSTRACT
Non‑alcoholic fatty liver disease (NAFLD) is a manifestation of metabolic syndrome in the liver and is closely associated with diabetes; however, its pathogenesis remains to be elucidated. Carbohydrate responsive element binding protein (ChREBP), the hub of glucolipid metabolism, regulates the induction of fatty acid synthase (FASN), the key enzyme of de novo lipogenesis, by directly binding to carbohydrate response element (ChoRE) in its promoter. Investigations of histone modifications on NAFLD remain in their infancy. In the present study, by using ChIP, the association between histone modifications and FASN transcription was investigated and histone modifications in FASN modulated by ChREBP were measured. It was demonstrated that ChREBP induced FASN ChREBP‑ChoRE binding to accelerate the expression of FASN, leading to hepatocellular steatosis by facilitating H3 and H4 acetylation, H3K4 trimethylation and the phosphorylation of H3S10, but inhibiting the trimethylation of H3K9 and H4K20 in FASN promoter regions of HepG2 and L02 cells. It was also found that ChREBP‑ChoRE binding of FASN relied on histone acetylation and that the transcriptional activity of ChREBP on FASN is required, based on the premise that histone acetylation causes conformational changes in FASN chromatin. This indicated histone acetylation as a crucial mechanism involved in the transcription of FASN modulated by ChREBP. Consequently, the present study provides further insight into the pathophysiology and a novel therapeutic potential of NAFLD based on epigenetic mechanisms.
Sigma-Aldrich
MISSION® esiRNA, targeting human MLXIPL
Sigma-Aldrich
Anti-trimethyl-Histone H4 (Lys20) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone MC463, rabbit monoclonal, culture supernatant, clone MC463, Upstate®