Merck
  • Salt-Responsive Metabolite, β-Hydroxybutyrate, Attenuates Hypertension.
Salt-Responsive Metabolite, β-Hydroxybutyrate, Attenuates Hypertension.
Cell reports (2018-10-18)
Saroj Chakraborty, Sarah Galla, Xi Cheng, Ji-Youn Yeo, Blair Mell, Vishal Singh, BengSan Yeoh, Piu Saha, Anna V Mathew, Matam Vijay-Kumar, Bina Joe
ABSTRACT
Dietary salt reduction and exercise are lifestyle modifications for salt-sensitive hypertensives. While exercise has prominent metabolic effects, salt has an adverse effect on metabolic syndrome, of which hypertension is a hallmark. We hypothesized that dietary salt impacts metabolism in a salt-sensitive model of hypertension. An untargeted metabolomic approach demonstrates lower circulating levels of the ketone body, beta-hydroxybutyrate (βOHB), in high salt-fed hypertensive rats. Despite the high salt intake, specific rescue of βOHB levels by nutritional supplementation of its precursor, 1,3-butanediol, attenuates hypertension and protects kidney function. This beneficial effect of βOHB was likely independent of gut-microbiotal and Th17-mediated effects of salt and instead facilitated by βOHB inhibiting the renal Nlrp3 inflammasome. The juxtaposed effects of dietary salt and exercise on salt-sensitive hypertension, which decrease and increase βOHB respectively, indicate that nutritional supplementation of a precursor of βOHB provides a similar benefit to salt-sensitive hypertension as exercise.
Sigma-Aldrich
(±)-1,3-Butanediol, ReagentPlus®, 99.5%
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O128:B12, purified by gel-filtration chromatography
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 3,000-5,000