Merck
Spatiotemporal Proteomic Profiling of Human Cerebral Development.
Molecular & cellular proteomics : MCP (2017-07-09)
Ugljesa Djuric, Deivid C Rodrigues, Ihor Batruch, James Ellis, Patrick Shannon, Phedias Diamandis
ABSTRACT
Mass spectrometry (MS) analysis of human post-mortem central nervous system (CNS) tissue and induced pluripotent stem cell (iPSC)-based directed differentiations offer complementary avenues to define protein signatures of neurodevelopment. Methodological improvements of formalin-fixed, paraffin-embedded (FFPE) protein isolation now enable widespread proteomic analysis of well-annotated archival tissue samples in the context of development and disease. Here, we utilize a shotgun label-free quantification (LFQ) MS method to profile magnetically enriched human cortical neurons and neural progenitor cells (NPCs) derived from iPSCs. We use these signatures to help define spatiotemporal protein dynamics of developing human FFPE cerebral regions. We show that the use of high resolution Q Exactive mass spectrometers now allow simultaneous quantification of >2700 proteins in a single LFQ experiment and provide sufficient coverage to define novel biomarkers and signatures of NPC maintenance and differentiation. Importantly, we show that this abbreviated strategy allows efficient recovery of novel cytoplasmic, membrane-specific and synaptic proteins that are shared between both in vivo and in vitro neuronal differentiation. This study highlights the discovery potential of non-comprehensive high-throughput proteomic profiling of unfractionated clinically well-annotated FFPE human tissue from a diverse array of development and diseased states.
Sigma-Aldrich
Anti-CRABP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FLNC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution