Merck
Structure of outer membrane protein G by solution NMR spectroscopy.
Proceedings of the National Academy of Sciences of the United States of America (2007-10-04)
Binyong Liang, Lukas K Tamm
ABSTRACT
The bacterial outer membrane protein G (OmpG), a monomeric pH-gated porin, was overexpressed in Escherichia coli and refolded in beta-octyl glucoside micelles. After transfer into dodecylphosphocholine micelles, the solution structure of OmpG was determined by solution NMR spectroscopy at pH 6.3. Complete backbone assignments were obtained for 234 of 280 residues based on CA, CB, and CO connection pathways determined from a series of TROSY-based 3D experiments at 800 MHz. The global fold of the 14-stranded beta-barrel was determined based on 133 long-range NOEs observed between neighboring strands and local chemical shift and NOE information. The structure of the barrel is very similar to previous crystal structures, but the loops of the solution structure are quite flexible.
Product Description
Avanti
MAPCHO-10, Avanti Research - A Croda Brand 850334P, powder
Avanti
MAPCHO-12, Avanti Research - A Croda Brand