Merck
  • Syringic acid prevents skin carcinogenesis via regulation of NoX and EGFR signaling.
Syringic acid prevents skin carcinogenesis via regulation of NoX and EGFR signaling.
Biochemical pharmacology (2018-06-11)
Su Jeong Ha, Jangho Lee, Joon Park, Young Ho Kim, Nam Hyouck Lee, Young Eon Kim, Kyung-Mo Song, Pahn-Shick Chang, Chul-Ho Jeong, Sung Keun Jung
ABSTRACT
Validation of nutraceutical and pharmaceutical targets is essential for the prediction of physiological and side effects. Epidemiologic evidence and molecular studies suggest that non-melanoma skin cancer is directly associated with excessive exposure to ultraviolet (UV) radiation. The aim of the present study was to evaluate the inhibitory effects of syringic acid on UVB-induced signaling and skin carcinogenesis, and determine the molecular targets. Treatment of human epidermal keratinocytes (HaCaT) cells with syringic acid resulted in the suppression of UVB-induced cyclooxygenase-2, matrix metalloproteinase-1, and prostaglandin E2 expression as well as activator protein-1 activity. Moreover, syringic acid inhibited the UVB-induced phosphorylation of mitogen-activated protein kinases and Akt signaling pathways as well as epidermal growth factor receptor (EGFR). Syringic acid treatment further inhibited intracellular reactive oxygen species and protein-tyrosine phosphatase-κ activity, a regulator of EGFR activation. Syringic acid and the antioxidant N-acetyl-l-cysteine inhibited UVB-induced nicotinamide adenine dinucleotide phosphate (NADPH) oxidase activity. In vivo, pretreatment of mouse skin with syringic acid significantly suppressed UVB-induced skin tumor incidence in a dose-dependent manner. Overall, these results indicate that syringic acid exerts potent chemopreventive activity in skin carcinogenesis mainly by inhibition of the Nox/PTP-κ/EGFR axis. Syringic acid might serve as an effective chemopreventive and therapeutic agent against UVB-mediated skin cancer.
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Pricing and availability is not currently available.
Supelco
Syringic acid, analytical standard
Pricing and availability is not currently available.