Merck
  • Granulocyte-macrophage colony-stimulating factor neuroprotective activities in Alzheimer's disease mice.
Granulocyte-macrophage colony-stimulating factor neuroprotective activities in Alzheimer's disease mice.
Journal of neuroimmunology (2018-03-27)
Tomomi Kiyota, Jatin Machhi, Yaman Lu, Bhagyalaxmi Dyavarshetty, Maryam Nemati, Izumi Yokoyama, R L Mosley, Howard E Gendelman
ABSTRACT
We investigated the effects of granulocyte-macrophage colony stimulating factor (GM-CSF) on behavioral and pathological outcomes in Alzheimer's disease (AD) and non-transgenic mice. GM-CSF treatment in AD mice reduced brain amyloidosis, increased plasma Aβ, and rescued cognitive impairment with increased hippocampal expression of calbindin and synaptophysin and increased levels of doublecortin-positive cells in the dentate gyrus. These data extend GM-CSF pleiotropic neuroprotection mechanisms in AD and include regulatory T cell-mediated immunomodulation of microglial function, Aβ clearance, maintenance of synaptic integrity, and induction of neurogenesis. Together these data support further development of GM-CSF as a neuroprotective agent for AD.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-LRP, Light Chain Mouse mAb (5A6), liquid, clone 5A6, Calbiochem®