Merck
Interactions between the human RNA polymerase II subunits.
The Journal of biological chemistry (1997-07-04)
J Acker, M de Graaff, I Cheynel, V Khazak, C Kedinger, M Vigneron
ABSTRACT
As an initial approach to characterizing the molecular structure of the human RNA polymerase II (hRPB), we systematically investigated the protein-protein contacts that the subunits of this enzyme may establish with each other. To this end, we applied a glutathione S-transferase-pulldown assay to extracts from Sf9 insect cells, which were coinfected with all possible combinations of recombinant baculoviruses expressing hRPB subunits, either as untagged polypeptides or as glutathione S-transferase fusion proteins. This is the first comprehensive study of interactions between eukaryotic RNA polymerase subunits; among the 116 combinations of hRPB subunits tested, 56 showed significant to strong interactions, whereas 60 were negative. Within the intricate network of interactions, subunits hRPB3 and hRPB5 play a central role in polymerase organization. These subunits, which are able to homodimerize and to interact, may constitute the nucleation center for polymerase assembly, by providing a large interface to most of the other subunits.
Sigma-Aldrich
RNA Polymerase II, p33 subunit, GST tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)